ADVH49B82391 AChemBlock

3-(2,4-difluorophenyl)azetidin-3-ol

  • Cas Number : 1388073-15-1
  • Molecular Weight : 185.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-2-4-difluorophenyl-azetidin-3-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80317
Availability details for
Quantity
  • Availability
  • Add to Cart