ADVH49B82392 AChemBlock

(2S)-4-benzyloxycarbonyl-1-tert-butoxycarbonyl-piperazine-2-carboxylic acid

  • Cas Number : 138775-03-8
  • Molecular Weight : 364.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2s-4-benzyloxycarbonyl-1-tert-butoxycarbonyl-piperazine-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80323
Availability details for
Quantity
  • Availability
  • Add to Cart