ADVH49B82393 AChemBlock

3-aminooxetane-3-carboxamide

  • Cas Number : 138650-60-9
  • Molecular Weight : 116.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-aminooxetane-3-carboxamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80328
Availability details for
Quantity
  • Availability
  • Add to Cart