ADVH49B82395 AChemBlock

3-(4-chlorophenyl)cyclobutane-1-carboxylic acid

  • Cas Number : 1384429-60-0
  • Molecular Weight : 210.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-4-chlorophenyl-cyclobutane-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80346
Availability details for
Quantity
  • Availability
  • Add to Cart