ADVH49B82396 AChemBlock

5-(2-amino-4-oxo-3-(2-(trifluoromethyl)phenyl)-3,4-dihydroquinazolin-6-yl)-N-(2,4-difluorophenyl)-2-methoxypyridine-3-sulfonamide

  • Cas Number : 1384097-27-1
  • Molecular Weight : 603.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/5-2-amino-4-oxo-3-2-trifluoromethyl-phenyl-3-4-dihydroquinazolin-6-yl-n-2-4-difluorophenyl-2-methoxypyridine-3-sulfonamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86604
Availability details for
Quantity
  • Availability
  • Add to Cart