ADVH49B82397 AChemBlock

(E)-2-(2-(pyridin-4-yl)vinyl)aniline

  • Cas Number : 138386-71-7
  • Molecular Weight : 196.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/e-2-2-pyridin-4-yl-vinyl-aniline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86786
Availability details for
Quantity
  • Availability
  • Add to Cart