ADVH49B82399 AChemBlock

5-bromo-2-(trifluoromethyl)pyrimidin-4-amine

  • Cas Number : 1379352-61-0
  • Molecular Weight : 242

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-trifluoromethyl-pyrimidin-4-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80381
Availability details for
Quantity
  • Availability
  • Add to Cart