ADVH49B8239C AChemBlock

3-bromo-1,4-dimethyl-pyrazole

  • Cas Number : 13745-59-0
  • Molecular Weight : 175.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-1-4-dimethyl-pyrazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80441
Availability details for
Quantity
  • Availability
  • Add to Cart