ADVH49B8239F AChemBlock

[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridyl]methanamine dihydrochloride

  • Cas Number : 1374451-84-9
  • Molecular Weight : 307.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-2-pyridyl-methanamine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80443
Availability details for
Quantity
  • Availability
  • Add to Cart