ADVH49B823A1 AChemBlock

2-bromoimidazo[2,1-b][1,3,4]thiadiazole-6-carboxylic acid

  • Cas Number : 1373253-24-7
  • Molecular Weight : 248.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-bromoimidazo-2-1-b-1-3-4-thiadiazole-6-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86545
Availability details for
Quantity
  • Availability
  • Add to Cart