ADVH49B823A3 AChemBlock

2--Methyl-4-(hydroxymethyl)phenylboronic acid pinacol ester

  • Cas Number : 1370732-71-0
  • Molecular Weight : 266.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-4-hydroxymethyl-phenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70549
Availability details for
Quantity
  • Availability
  • Add to Cart