ADVH49B823A5 AChemBlock

5-fluoro-3-nitropyridin-2-ol

  • Cas Number : 136888-20-5
  • Molecular Weight : 158.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-fluoro-3-nitropyridin-2-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71712
Availability details for
Quantity
  • Availability
  • Add to Cart