ADVH49B823A6 AChemBlock

2H-[1,3]dioxolo[4,5-b]pyridin-5-amine

  • Cas Number : 1368345-12-3
  • Molecular Weight : 138.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2h-1-3-dioxolo-4-5-b-pyridin-5-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80519
Availability details for
Quantity
  • Availability
  • Add to Cart