ADVH49B823AD AChemBlock

4-bromo-2-(trifluoromethoxy)pyridine

  • Cas Number : 1361880-72-9
  • Molecular Weight : 242

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-2-trifluoromethoxy-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80630
Availability details for
Quantity
  • Availability
  • Add to Cart