ADVH49B823B2 AChemBlock

3,3-dimethyltetrahydropyran-4-amine

  • Cas Number : 1357396-54-3
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-3-dimethyltetrahydropyran-4-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80663
Availability details for
Quantity
  • Availability
  • Add to Cart