ADVH49B823B7 AChemBlock

(4-amino-2-pyridyl)methanol hydrochloride

  • Cas Number : 1354940-95-6
  • Molecular Weight : 160.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-amino-2-pyridyl-methanol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80709
Availability details for
Quantity
  • Availability
  • Add to Cart