ADVH49B823B9 AChemBlock

RP-54745

  • Cas Number : 135330-08-4
  • Molecular Weight : 297.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/rp-54745.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86717
Availability details for
Quantity
  • Availability
  • Add to Cart