ADVH49B823BA AChemBlock

4-fluoropyrazolo[1,5-a]pyridine-3-carboxylic acid

  • Cas Number : 1352625-33-2
  • Molecular Weight : 180.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-fluoropyrazolo-1-5-a-pyridine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80789
Availability details for
Quantity
  • Availability
  • Add to Cart