ADVH49B823BC AChemBlock

methyl 3-fluoro-1H-pyrrolo[2,3-b]pyridine-4-carboxylate

  • Cas Number : 1352397-87-5
  • Molecular Weight : 194.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-fluoro-1h-pyrrolo-2-3-b-pyridine-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80811
Availability details for
Quantity
  • Availability
  • Add to Cart