ADVH49B823BD AChemBlock

methyl 4-bromo-3-chloro-1H-indole-6-carboxylate

  • Cas Number : 1352393-64-6
  • Molecular Weight : 288.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-bromo-3-chloro-1h-indole-6-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80820
Availability details for
Quantity
  • Availability
  • Add to Cart