ADVH49B823BE AChemBlock

(1,1-dioxothietan-2-yl)methanamine

  • Cas Number : 1352242-96-6
  • Molecular Weight : 135.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-1-dioxothietan-2-yl-methanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80822
Availability details for
Quantity
  • Availability
  • Add to Cart