ADVH49B823C0 AChemBlock

6-bromo-5-methoxy-1H-pyrazolo[4,3-b]pyridine

  • Cas Number : 1351813-60-9
  • Molecular Weight : 228.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-5-methoxy-1h-pyrazolo-4-3-b-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80827
Availability details for
Quantity
  • Availability
  • Add to Cart