ADVH49B823C2 AChemBlock

tert-butyl 4-(oxetan-3-ylamino)piperidine-1-carboxylate

  • Cas Number : 1349718-24-6
  • Molecular Weight : 256.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-oxetan-3-ylamino-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80843
Availability details for
Quantity
  • Availability
  • Add to Cart