ADVH49B823C4 AChemBlock

4-chloro-7-{7H-pyrrolo[2,3-d]pyrimidin-4-yl}-7H-pyrrolo[2,3-d]pyrimidine

  • Cas Number : 134965-85-8
  • Molecular Weight : 270.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-7-7h-pyrrolo-2-3-d-pyrimidin-4-yl-7h-pyrrolo-2-3-d-pyrimidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80844
Availability details for
Quantity
  • Availability
  • Add to Cart