ADVH49B823C5 AChemBlock

tert-butyl 3-(6-amino-3-pyridyl)azetidine-1-carboxylate

  • Cas Number : 1346673-86-6
  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-6-amino-3-pyridyl-azetidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80855
Availability details for
Quantity
  • Availability
  • Add to Cart