ADVH49B823C7 AChemBlock

6-bromo-5-methylpyrazolo[1,5-a]pyridine

  • Cas Number : 1345121-23-4
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-5-methylpyrazolo-1-5-a-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80870
Availability details for
Quantity
  • Availability
  • Add to Cart