ADVH49B823CC AChemBlock

3-Cyclohexyl-N-methoxy-N-methylpropanamide

  • Cas Number : 1342103-68-7
  • Molecular Weight : 199.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-cyclohexyl-n-methoxy-n-methylpropanamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70382
Availability details for
Quantity
  • Availability
  • Add to Cart