ADVH49B823CD AChemBlock

2-azaspiro[4.5]decane-1,8-dione

  • Cas Number : 1341037-14-6
  • Molecular Weight : 167.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-azaspiro-4-5-decane-1-8-dione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80955
Availability details for
Quantity
  • Availability
  • Add to Cart