ADVH49B823CE AChemBlock

tert-butyl 3-(aminomethyl)-1-oxa-7-azaspiro[4.4]nonane-7-carboxylate

  • Cas Number : 1341035-92-4
  • Molecular Weight : 256.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-aminomethyl-1-oxa-7-azaspiro-4-4-nonane-7-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80956
Availability details for
Quantity
  • Availability
  • Add to Cart