ADVH49B823CF AChemBlock

methyl 5-(pyridin-2-yl)-1H-pyrazole-3-carboxylate

  • Cas Number : 1340553-59-4
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-pyridin-2-yl-1h-pyrazole-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86534
Availability details for
Quantity
  • Availability
  • Add to Cart