ADVH49B823D4 AChemBlock

3-chloroisoquinoline-5-carboxylic acid

  • Cas Number : 1337880-37-1
  • Molecular Weight : 207.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-chloroisoquinoline-5-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S81004
Availability details for
Quantity
  • Availability
  • Add to Cart