ADVH49B823D9 AChemBlock

BP-1-102

  • Cas Number : 1334493-07-0
  • Molecular Weight : 626.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/bp-1-102.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72381
Availability details for
Quantity
  • Availability
  • Add to Cart