ADVH49B823DF AChemBlock

CZC-25146

  • Cas Number : 1330003-04-7
  • Molecular Weight : 488.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/czc-25146.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86746
Availability details for
Quantity
  • Availability
  • Add to Cart