ADVH49B823E0 AChemBlock

ethanesulfonic anhydride

  • Cas Number : 13223-06-8
  • Molecular Weight : 202.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethanesulfonic-anhydride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71739
Availability details for
Quantity
  • Availability
  • Add to Cart