ADVH49B823E1 AChemBlock

7-bromo-1,2,3,4-tetrahydronaphthalen-2-one

  • Cas Number : 132095-54-6
  • Molecular Weight : 225.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-1-2-3-4-tetrahydronaphthalen-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S81102
Availability details for
Quantity
  • Availability
  • Add to Cart