ADVH49B823E3 AChemBlock

4-bromopyridine-2-carbaldehyde

  • Cas Number : 131747-63-2
  • Molecular Weight : 186.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-bromopyridine-2-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71742
Availability details for
Quantity
  • Availability
  • Add to Cart