ADVH49B823EA AChemBlock

2-(fluoromethyl)cyclopropane-1-carboxylic acid

  • Cas Number : 1314960-53-6
  • Molecular Weight : 118.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-fluoromethyl-cyclopropane-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S81153
Availability details for
Quantity
  • Availability
  • Add to Cart