ADVH49B823F0 AChemBlock

(4-Amino-3-methoxyphenyl)(4-(4-methylpiperazin-1-yl)piperidin-1-yl)methanone

  • Cas Number : 1309887-09-9
  • Molecular Weight : 332.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-amino-3-methoxyphenyl-4-4-methylpiperazin-1-yl-piperidin-1-yl-methanone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86715
Availability details for
Quantity
  • Availability
  • Add to Cart