ADVH49B823F3 AChemBlock

tert-butyl N-[(1S)-4-[5-(3-cyano-4-isopropoxy-phenyl)-1,2,4-oxadiazol-3-yl]indan-1-yl]carbamate

  • Cas Number : 1306763-71-2
  • Molecular Weight : 460.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-1s-4-5-3-cyano-4-isopropoxy-phenyl-1-2-4-oxadiazol-3-yl-indan-1-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S81251
Availability details for
Quantity
  • Availability
  • Add to Cart