ADVH49B823F4 AChemBlock

tert-butyl 3-(chloromethyl)-6,7-dihydro-4H-triazolo[1,5-a]pyrazine-5-carboxylate

  • Cas Number : 1305336-27-9
  • Molecular Weight : 272.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-chloromethyl-6-7-dihydro-4h-triazolo-1-5-a-pyrazine-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S81265
Availability details for
Quantity
  • Availability
  • Add to Cart