ADVH49B823F6 AChemBlock

2-chloro-4-phenylpyrimidine

  • Cas Number : 13036-50-5
  • Molecular Weight : 190.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-4-phenylpyrimidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71748
Availability details for
Quantity
  • Availability
  • Add to Cart