ADVH49B823FC AChemBlock

4-fluoro-2-(2H-1,2,3-triazol-2-yl)benzoic acid

  • Cas Number : 1293284-50-0
  • Molecular Weight : 207.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-fluoro-2-2h-1-2-3-triazol-2-yl-benzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86855
Availability details for
Quantity
  • Availability
  • Add to Cart