ADVH49B82400 AChemBlock

5-amino-2-bromo-pyridine-4-carbaldehyde

  • Cas Number : 1289021-84-6
  • Molecular Weight : 201.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-amino-2-bromo-pyridine-4-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S81346
Availability details for
Quantity
  • Availability
  • Add to Cart