ADVH49B82401 AChemBlock

1-ethoxycyclopropanecarboxylic acid

  • Cas Number : 128732-53-6
  • Molecular Weight : 130.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-ethoxycyclopropanecarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S81353
Availability details for
Quantity
  • Availability
  • Add to Cart