ADVH49B8240A AChemBlock

2-(3-bromophenyl)azetidine

  • Cas Number : 1270556-17-6
  • Molecular Weight : 212.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-3-bromophenyl-azetidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S81418
Availability details for
Quantity
  • Availability
  • Add to Cart