ADVH49B8240D AChemBlock

1-(3-chloro-2-pyridyl)ethanol

  • Cas Number : 1269430-33-2
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-3-chloro-2-pyridyl-ethanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S81437
Availability details for
Quantity
  • Availability
  • Add to Cart