ADVH49B82417 AChemBlock

2-(bromomethyl)-1-methyl-3-(trifluoromethyl)benzene

  • Cas Number : 1261683-90-2
  • Molecular Weight : 253.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-bromomethyl-1-methyl-3-trifluoromethyl-benzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S81517
Availability details for
Quantity
  • Availability
  • Add to Cart