ADVH49B8241F AChemBlock

tert-butyl 3-(6-hydroxy-2-methylpyrimidin-4-yl)piperidine-1-carboxylate

  • Cas Number : 1260178-75-3
  • Molecular Weight : 293.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-6-hydroxy-2-methylpyrimidin-4-yl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86543
Availability details for
Quantity
  • Availability
  • Add to Cart