ADVH49B82420 AChemBlock

6-bromo-3,3-dimethyl-1H,2H,3H-pyrrolo[3,2-b]pyridine

  • Cas Number : 1259512-11-2
  • Molecular Weight : 227.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-3-3-dimethyl-1h-2h-3h-pyrrolo-3-2-b-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S81603
Availability details for
Quantity
  • Availability
  • Add to Cart