ADVH49B82421 AChemBlock

3-(tosyloxy)propionic acid

  • Cas Number : 1259387-78-4
  • Molecular Weight : 244.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-tosyloxy-propionic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86737
Availability details for
Quantity
  • Availability
  • Add to Cart